Titanium ≥99.98% (metals basis), wire, Ø 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals
Warning

00362-G1 00362-G5
CAAA00362-G1EA 221.2 CAD
CAAA00362-G1 CAAA00362-G5
Titanium ≥99.98% (metals basis), wire, Ø 0.5 mm (0.02 in)
Titanium

MDL: MFCD00011264


Purity: 99.98% CAS No.: 7440-32-6 Physical Form: ≈0.88g/m Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR