Titanium ≥99.98% (metals basis), wire, Ø 0.75 mm (0.03 in)
Supplier: Thermo Scientific Chemicals
|
Warning
|
00361-BU
00361-BY
CAAA00361-BUEA
99.07
CAD
CAAA00361-BU
CAAA00361-BY
Titanium ≥99.98% (metals basis), wire, Ø 0.75 mm (0.03 in)
Titanium
MDL: MFCD00011264
Purity: 99.98%
CAS No.: 7440-32-6
Physical Form: ≈1.99g/m, Temper: as drawn
Odor: Odorless
MDL No.: MFCD00011264
Formula:
Ti Boiling Pt: 3262 °C (1013 hPa) Melting Pt: 1660 °C Density: 4.51 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 Merck Index: 12,09610 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...