Tin ≥99.998% (metals basis), foil, Thickness: 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

00344-GE 00344-GU
CAAA00344-GEEA 221.2 CAD
CAAA00344-GE CAAA00344-GU
Tin ≥99.998% (metals basis), foil, Thickness: 0.127 mm (0.005 in)
Tin

Purity: 99.998% CAS No.: 7440-31-5 Physical Form: ≈6.9g/75x100mm Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR