Tantalum ≥99.6% (metals basis), powder -325 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
00337-14
00337-22
CAAA00337-14EA
254.38
CAD
CAAA00337-14
CAAA00337-22
Tantalum ≥99.6% (metals basis), powder -325 mesh
Tantalum
MDL: MFCD00011252
Notes: 99.6% purity includes H, N, C, O
Purity: 99.9%
CAS No.: 7440-25-7
Appearance: Black
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00011252
Formula:
Ta Density: 16,6 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 EINECS: 231-135-5 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...