Tantalum ≥99.997% (metals basis), foil, Thickness: 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals

00335-FF 00335-FQ
CAAA00335-FFEA 175.38 CAD
CAAA00335-FF CAAA00335-FQ
Tantalum ≥99.997% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Tantalum

MDL: MFCD00011252


Purity: 99.997% CAS No.: 7440-25-7 Physical Form: ≈0.26g/25x25mm Odor: Odorless MDL No.: MFCD00011252

Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR