Tantalum hard ≥99.95% (metals basis), foil, hard, Thickness: 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

00331-GE 00331-GU
CAAA00331-GEEA 284.4 CAD
CAAA00331-GE CAAA00331-GU
Tantalum hard ≥99.95% (metals basis), foil, hard, Thickness: 0.127 mm (0.005 in)
Tantalum

MDL: MFCD00011252


Purity: 99.95% CAS No.: 7440-25-7 Physical Form: ≈15.8g/75x100mm, Temper: as rolled Appearance: Grey-black Odor: Odorless MDL No.: MFCD00011252

Formula: Ta
Boiling Pt: 5427 °C (1013 hPa)
Melting Pt: 2996 °C
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR