Terbium ≥99.9% (REO, rare earth oxide basis), pieces

Supplier: Thermo Scientific Chemicals
Danger

00324-03 00324-06
CAAA00324-03EA 126.72 CAD
CAAA00324-03 CAAA00324-06
Terbium ≥99.9% (REO, rare earth oxide basis), pieces
Terbium

MDL: MFCD00011256


Purity: 99.9% CAS No.: 7440-27-9 Physical Form: Packaged under argon Appearance: Silver-grey Odor: Odorless UN No.: 3178 MDL No.: MFCD00011256

Formula: Tb
Boiling Pt: 3041 °C (1013 hPa)
Melting Pt: 1356 °C
Density: 8.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011256
CAS Number: 7440-27-9
EINECS: 231-137-6
UN: 3178
ADR: 4.1,II
Merck Index: 12,09300

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR