Terbium ≥99.9% (REO, rare earth oxide basis), ingot (max. 0,4% Ta)

Supplier: Thermo Scientific Chemicals

00320-04 00320-09
CAAA00320-04EA 169.06 CAD
CAAA00320-04 CAAA00320-09
Terbium ≥99.9% (REO, rare earth oxide basis), ingot (max. 0,4% Ta)
Terbium

MDL: MFCD00011256


Purity: 99.9% CAS No.: 7440-27-9 Physical Form: Packaged in air-stable poly bottle MDL No.: MFCD00011256

Formula: Tb
Boiling Pt: 3041 °C (1013 hPa)
Melting Pt: 1356 °C
Density: 8.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011256
CAS Number: 7440-27-9
EINECS: 231-137-6
Merck Index: 12,09300

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR