Silver 99,9 Metals basis, wire, annealed, Ø 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

00303-G5 00303-G9 00303-H4
CAAA00303-G5EA 80.58 CAD
CAAA00303-G5 CAAA00303-G9 CAAA00303-H4
Silver 99,9 Metals basis, wire, annealed, Ø 0.127 mm (0.005 in)
Silver

MDL: MFCD00003397


Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈0.13g/m, Temper: soft Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR