Samarium ≥99.9% (REO, rare earth oxide basis), ingot

Supplier: Thermo Scientific Chemicals

00295-14 00295-22
CAAA00295-14EA 210.14 CAD
CAAA00295-14 CAAA00295-22
Samarium ≥99.9% (REO, rare earth oxide basis), ingot
Samarium

MDL: MFCD00011233


Purity: 99.9% CAS No.: 7440-19-9 Physical Form: Packaged in air stable poly bottle Odor: Odorless MDL No.: MFCD00011233

Formula: Sm
Boiling Pt: 1900 °C (1013 hPa)
Melting Pt: 1076.85 °C
Density: 7.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011233
CAS Number: 7440-19-9
EINECS: 231-128-7
Merck Index: 12,08498

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR