Scandium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.127 mm (0.005 in)
Supplier: Thermo Scientific Chemicals
00294-FF
00294-FI
CAAA00294-FFEA
483.48
CAD
CAAA00294-FF
CAAA00294-FI
Scandium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.127 mm (0.005 in)
Scandium
MDL: MFCD00016323
Purity: 99.9%
CAS No.: 7440-20-2
Physical Form: ≈0.24g/25x25mm, Packaged under argon
Odor: Odorless
MDL No.: MFCD00016323
Formula:
Sc Boiling Pt: 2730 °C (1013 hPa) Melting Pt: 1541 °C Density: 2.989 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016323 CAS Number: 7440-20-2 EINECS: 231-129-2 Merck Index: 12,08537 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...