Praseodymium 99% REO basis, ingot
Supplier: Thermo Scientific Chemicals
|
Danger
|
00267-09
00267-18
CAAA00267-09EA
170.64
CAD
CAAA00267-09
CAAA00267-18
Praseodymium 99% REO basis, ingot
Praseodymium
MDL: MFCD00011174
Purity: 99.9%
CAS No.: 7440-10-0
Physical Form: Packaged under mineral oil
Sensitivity: Air and Moisture Sensitive
Odor: Odorless
UN No.: 3178
MDL No.: MFCD00011174
Formula:
Pr MW: 140.91 Storage Temperature: Ambient |
MDL Number:
MFCD00011174 CAS Number: 7440-10-0 EINECS: 231-120-3 UN: 3178 ADR: 4.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...