Niobium ≥99.97% (metals basis excluding Ta), foil (max. 0.06% Ta), Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

00238-FF 00238-FQ
CAAA00238-FFEA 76.63 CAD
CAAA00238-FF CAAA00238-FQ
Niobium ≥99.97% (metals basis excluding Ta), foil (max. 0.06% Ta), Thickness: 0.25 mm (0.01 in)
Niobium

MDL: MFCD00011126


Purity: 99.97% CAS No.: 7440-03-1 Physical Form: ≈1.34g/25x25mm Appearance: Grey or silver Odor: Odorless MDL No.: MFCD00011126

Formula: Nb
Boiling Pt: 3700 °C (1013 hPa)
Melting Pt: 2468 °C
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR