Niobium ≥99.97% (metals basis excluding Ta), foil (max. 0.06% Ta), Thickness: 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

00236-FF 00236-FQ
CAAA00236-FFEA 59.57 CAD
CAAA00236-FF CAAA00236-FQ
Niobium ≥99.97% (metals basis excluding Ta), foil (max. 0.06% Ta), Thickness: 0.127 mm (0.005 in)
Niobium

MDL: MFCD00011126


Purity: 99.97% CAS No.: 7440-03-1 Physical Form: ≈0.68g/25x25mm Appearance: Grey or silver Odor: Odorless MDL No.: MFCD00011126

Formula: Nb
Boiling Pt: 3700 °C (1013 hPa)
Melting Pt: 2468 °C
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR