Nickel ≥99.5% (metals basis), foil, annealed, Thickness: 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals
Danger

00228-FT 00228-FW
CAAA00228-FTEA 139.99 CAD
CAAA00228-FT CAAA00228-FW
Nickel ≥99.5% (metals basis), foil, annealed, Thickness: 0.025 mm (0.001 in)
Nickel

MDL: MFCD00011137


Purity: 99.5% CAS No.: 7440-02-0 Physical Form: ≈1.4g/25x250mm Odor: Odorless MDL No.: MFCD00011137

Formula: Ni
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1453 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR