Nickel ≥99% (metals basis excluding Co), powder -100 mesh (max. 0.1% Co)
Supplier: Thermo Scientific Chemicals
|
Danger
|
00224-14
00224-30
00224-A1
CAAA00224-14EA
35.87
CAD
CAAA00224-14
CAAA00224-30
CAAA00224-A1
Nickel ≥99% (metals basis excluding Co), powder -100 mesh (max. 0.1% Co)
Nickel
MDL: MFCD00011137
Purity: 99+%
CAS No.: 7440-02-0
Appearance: Silvery
Odor: Odorless
MDL No.: MFCD00011137
Formula:
Ni Boiling Pt: 2371 °C (1013 hPa) Melting Pt: 1455 °C Density: 8.9 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...