Neodymium ≥99.1% (metals basis), ingot

Supplier: Thermo Scientific Chemicals

00214-14 00214-22
CAAA00214-14EA 202.24 CAD
CAAA00214-14 CAAA00214-22
Neodymium ≥99.1% (metals basis), ingot
Neodymium

MDL: MFCD00011130


Purity: 99.1% CAS No.: 7440-00-8 Physical Form: Packaged under mineral oil Appearance: Grey/silver MDL No.: MFCD00011130

Formula: Nd
MW: 144.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011130
CAS Number: 7440-00-8
EINECS: 231-109-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR