Iron ≥99.5% (metals basis), spherical powder <10 µm

Supplier: Thermo Scientific Chemicals
Danger

00170-14 00170-30 00170-A1
CAAA00170-14EA 95.12 CAD
CAAA00170-14 CAAA00170-30 CAAA00170-A1
Iron ≥99.5% (metals basis), spherical powder <10 µm
Iron

MDL: MFCD00010999


Purity: 99.9+% CAS No.: 7439-89-6 Appearance: Grey to brown UN No.: 3089 MDL No.: MFCD00010999

Formula: Fe
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1536 °C
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR