Indium ≥99.999% (metals basis), powder, Puratronic® -325 mesh

Supplier: Thermo Scientific Chemicals
Warning

00151-06 00151-14
CAAA00151-06EA 273.34 CAD
CAAA00151-06 CAAA00151-14
Indium ≥99.999% (metals basis), powder, Puratronic® -325 mesh
Indium

MDL: MFCD00134048


Purity: 99.999% CAS No.: 7440-74-6 Molecular Formula: In Formula Weight: 114.82 Physical Form: Powder Density: 7.31 Appearance: Silver-grey Odor: Odorless Melting Point: 156.6° Boiling Point: 2072° UN No.: 3089 MDL No.: MFCD00134048

Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Density: 7.31
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
UN: 3089
ADR: 4.1 III
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR