Holmium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

00144-FF 00144-FI
CAAA00144-FFEA 404.48 CAD
CAAA00144-FF CAAA00144-FI
Holmium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.127 mm (0.005 in)
Holmium

MDL: MFCD00011049


Purity: 99.9% CAS No.: 7440-60-0 Physical Form: ≈0.70g/25x25mm, Packaged in mylar under argon Odor: Odorless MDL No.: MFCD00011049

Formula: Ho
Storage Temperature: Ambient
MDL Number: MFCD00011049
CAS Number: 7440-60-0
EINECS: 231-169-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR