Holmium ≥99.9% (metals basis excluding Ta), pieces

Supplier: Thermo Scientific Chemicals

00142-06 00142-14
CAAA00142-03EA 52.25 CAD
CAAA00142-03 CAAA00142-06 CAAA00142-14
Holmium ≥99.9% (metals basis excluding Ta), pieces
Holmium

MDL: MFCD00011049


Purity: 99.9% CAS No.: 7440-60-0 Physical Form: Packaged in air-stable poly bottle MDL No.: MFCD00011049

Formula: Ho
Storage Temperature: Ambient
MDL Number: MFCD00011049
CAS Number: 7440-60-0
EINECS: 231-169-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR