Gold ≥99.995% (metals basis), powder, Premion® -20/+100 mesh

Supplier: Thermo Scientific Chemicals

00136-03 00136-06
CAAA00136-03EA 459.78 CAD
CAAA00136-03 CAAA00136-06
Gold ≥99.995% (metals basis), powder, Premion® -20/+100 mesh
Gold

MDL: MFCD00003436


Purity: 99.995% CAS No.: 7440-57-5 Physical Form: Agglomerated MDL No.: MFCD00003436

Formula: Au
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR