Germanium ≥99.9999% (metals basis), pieces ≤ 2 cm (0.8 in)

Supplier: Thermo Scientific Chemicals

00123-09 00123-18
CAAA00123-09EA 342.86 CAD
CAAA00123-09 CAAA00123-18
Germanium ≥99.9999% (metals basis), pieces ≤ 2 cm (0.8 in)
Germanium

MDL: MFCD00085310


Purity: 99.9999% CAS No.: 7440-56-4 Physical Form: Electrical Resistivity: 40 ohm-cm Odor: Odorless MDL No.: MFCD00085310

Formula: Ge
MW: 72.61 g/mol
Boiling Pt: 2830 °C (1013 hPa)
Melting Pt: 937.2 °C
Density: 5.323 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085310
CAS Number: 7440-56-4
EINECS: 231-164-3
Merck Index: 12,04415

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR