Gadolinium ≥99.9% (REO, rare earth oxide basis), pieces ≤ 20 mm (0.79 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
00118-09
00118-18
CAAA00118-09EA
175.38
CAD
CAAA00118-09
CAAA00118-18
Gadolinium ≥99.9% (REO, rare earth oxide basis), pieces ≤ 20 mm (0.79 in)
Gadolinium
MDL: MFCD00011022
Purity: 99.9%
CAS No.: 7440-54-2
Physical Form: Packaged under argon in glass bottle
Appearance: Silver
UN No.: 3178
MDL No.: MFCD00011022
Formula:
Gd MW: 157.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011022 CAS Number: 7440-54-2 EINECS: 231-162-2 UN: 3178 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...