Dysprosium ≥99.9% (REO, rare earth oxide basis), pieces

Supplier: Thermo Scientific Chemicals
Danger

00106-09 00106-18
CAAA00106-09EA 229.1 CAD
CAAA00106-09 CAAA00106-18
Dysprosium ≥99.9% (REO, rare earth oxide basis), pieces
Dysprosium

MDL: MFCD00010982


Purity: 99.9% CAS No.: 7429-91-6 Physical Form: Packaged under argon Appearance: Silver-grey Odor: Odorless UN No.: 3178 MDL No.: MFCD00010982

Formula: Dy
MW: 162.5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010982
CAS Number: 7429-91-6
EINECS: 231-073-9
UN: 3178
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR