Dysprosium ≥99.8% (REO, rare earth oxide basis), ingot

Supplier: Thermo Scientific Chemicals

00102-14 00102-22
CAAA00102-14EA 186.44 CAD
CAAA00102-14 CAAA00102-22
Dysprosium ≥99.8% (REO, rare earth oxide basis), ingot
Dysprosium

MDL: MFCD00010982


Purity: 99.8% CAS No.: 7429-91-6 Physical Form: Packaged in air stable poly bottle Odor: Odorless MDL No.: MFCD00010982

Formula: Dy
Storage Temperature: Ambient
MDL Number: MFCD00010982
CAS Number: 7429-91-6
EINECS: 231-073-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR