Copper ≥99.999% (metals basis), wire, Puratronic® oxygen-free, Ø 0.64 mm (0.025 in)
Supplier: Thermo Scientific Chemicals
00098-G2
00098-G6
CAAA00098-G2EA
109.18
CAD
CAAA00098-G2
CAAA00098-G6
Copper ≥99.999% (metals basis), wire, Puratronic® oxygen-free, Ø 0.64 mm (0.025 in)
Copper
MDL: MFCD00010965
Purity: 99.999%
CAS No.: 7440-50-8
Physical Form: ≈2.87g/m
Odor: Odorless
MDL No.: MFCD00010965
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2595 Melting Pt: 1083 Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...