Cadmium ≥99.999% (metals basis), wire, Puratronic®, Ø 0.64 mm (0.025 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
00060-G4
00060-G7
CAAA00060-G4EA
164.32
CAD
CAAA00060-G4
CAAA00060-G7
Cadmium ≥99.999% (metals basis), wire, Puratronic®, Ø 0.64 mm (0.025 in)
Cadmium
MDL: MFCD00010914
Purity: 99.999%
CAS No.: 7440-43-9
Physical Form: ≈2.8g/m
MDL No.: MFCD00010914
Formula:
Cd Boiling Pt: 765 °C (1013 hPa) Melting Pt: 321 °C Density: 8.642 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010914 CAS Number: 7440-43-9 EINECS: 231-152-8 UN: 2570 ADR: 6.1,II Merck Index: 12,01649 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...