Aluminium ≥99.999% (metals basis), wire, Puratronic®, hard, Ø 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

00005-G5 00005-G9
CAAA00005-G5EA 92.27 CAD
CAAA00005-G5 CAAA00005-G9
Aluminium ≥99.999% (metals basis), wire, Puratronic®, hard, Ø 0.127 mm (0.005 in)
Aluminium

MDL: MFCD00134029


Purity: 99.999% CAS No.: 7429-90-5 Physical Form: ≈0.034g/m, Temper: as drawn Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR