Aluminium ≥99.9995% (metals basis), foil 50x125 mm (2.0x4.9 in), Thickness: 0.13 mm (0.005 in)
Supplier: Thermo Scientific Chemicals
00004-KF
00004-KI
CAAA00004-KFEA
116.6
CAD
CAAA00004-KF
CAAA00004-KI
Aluminium ≥99.9995% (metals basis), foil 50x125 mm (2.0x4.9 in), Thickness: 0.13 mm (0.005 in)
Aluminium
MDL: MFCD00134029
Purity: 99.9995%
CAS No.: 7429-90-5
Physical Form: ≈2.2g/50x125mm
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al Boiling Pt: 2327 °C (1013 hPa) Melting Pt: 660 °C Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...