PYREX® 47 mm Microfiltration Glassware Apparatus, Corning

Supplier: Corning
33970-47 33972-B
71001-592CS 560.21 CAD
71001-592 71001-598
PYREX® 47 mm Microfiltration Glassware Apparatus, Corning
Filters Bottle Top Filters
These microfiltration components are designed for the filtration of aqueous liquids. They may be used for the filtration of cell culture media, buffers or for particulate analysis of water samples.

  • Units have coarse porosity frits and accommodate 47 mm filters
  • All-glass design ensures chemical compatibility with most organic solvents
  • Components are interchangeable with competitive products

Ordering information: Heavy wall filtering flasks are also available on request (sold separately).

Delivery information: The unit includes 300 ml funnel, 47 mm fritted glass support base, aluminum clamp and #8 silicone stopper.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR