Heracell i CO₂ Incubators with Variable O₂ Control, Thermo Scientific
Supplier: Thermo Fisher Scientific
51026410 51026408 51026402 51026537 51026529 51026423 51026422 51026556 51026533 51026410 51026529 51026402 51026408 51026537 51026423 51026556 51026422 51032967 51032969 51032971 51032972 51032973 51032975 51032977
89187-188EA 26070 CAD
89187-188 89187-190 89187-192 89187-194 89187-196 89187-200 89187-202 89187-204 89187-206 CA97015-624 CA97015-626 CA97015-628 CA97015-632 CA97015-636 CA97015-656 CA97015-660 CA97015-664 76379-706 76379-708 76379-710 76379-712 76379-714 76379-716 76379-720
Heracell i CO₂ Incubators with Variable O₂ Control, Thermo Scientific
Incubators CO₂ Incubators
Heracell i CO₂ incubators with variable control of oxygen stimulate cell behaviors that are more predictive of the in vivo environment. Available in a choice of two ranges of O₂ control, 1 to 21% for physiological levels and 5 to 90% for hyperoxic studies.each is supplied with maintenance free zirconia oxide sensor is calibrated automatically and remains in place during the ContraCon high temperature decontamination, and a unique gas tight segmented inner door assembly enables access sections of the chamber without disturbing the entire environment to minimize recovery time, contaminationMore Product Information