Heracell i CO₂ Incubators, Thermo Scientific
Supplier: Thermo Fisher Scientific
51026282 51026406 51026331 51026420 51026282 51026406 51026331 50116048
89187-168EA 18486 CAD
89187-168 89187-172 89187-180 89187-184 CA97015-604 CA97015-608 CA97015-640 CA97015-606
Heracell i CO₂ Incubators, Thermo Scientific
Incubators CO₂ Incubators
Thermo Scientific Heracell i CO₂ incubators combines optimal culturing conditions with simplicity and ease of use.
Available in both stackable sizes (150 L and 240 L), incubators are available with a choice of CO₂ sensors. Standard thermal conductivity (TC) sensor features accurate gas sensor technology. Infrared (IR) sensor features exclusive split-beam technology for precise CO₂ control in fluctuating temperature and humidity conditions.
In-chamber sensors and probes with an integrated, heated humidity water reservoir facilitate fast recovery rates of all parameters after routine access. The exclusive iCAN touch screen controller provides complete data visibility to monitor all incubator interactions with on-screen menu prompts, error and usage logs, data logging and trend graphing, and multiple language selection. Heracell i CO₂ incubators feature the on-demand ContraCon 90° moist heat decontamination cycle that ensures worry-free cleaning and operation. The ContraCon on-demand decontamination has been proven effective by multiple third party testing labs against wide contaminants including bacteria, molds, fungal spores, and mycoplasma.
Certifications: FDA Class II Medical Device for IVF
CE Marked, CSA Approved.
Ordering information: Includes stainless steel inner casing, ContraCon decontamination, and iCAN touchscreen.