PYREX® Desiccators with 24/29 Standard Taper Stopcock, Vacuum Rated, Corning
Supplier: Corning
3121-150 3121-150CO 3121-200 3121-200CO 3121-250 3121-STPK
89134-402CS 838.84 CAD
89134-402 89134-404 89134-406 89134-408 89134-410 89134-414
PYREX® Desiccators with 24/29 Standard Taper Stopcock, Vacuum Rated, Corning
Desiccators Desiccators, Glass
These PYREX® desiccators have a rugged 24/29 standard tapered stopcock with a tooled hose connection that serves as the vacuum inlet.
  • The tubulation Ext.Ø is approximately 10 mm
  • Material is Borosilicate Glass Code 7740
  • This design eliminates the need for grease
It is designed so that its manipulation is not affected by the vacuum. To introduce vacuum into the desiccator, loosen the stopcock to disengage the PTFE plug from contact with the glass. A half turn from this position closes the desiccator.
Replacement covers and stopcock are listed in the Order tab below. The recommended replacement lower bowls which are interchangeable with these desiccators, are as follows: for the 2.4 L desiccator, use replacement bowl Cat. No. 89134-388; for the 5.8 L desiccator, use replacement bowl Cat. No. 89134-394; and for the 10.5 L desiccator, use replacement bowl Cat. No. 89134-400.
Ordering information: Plate is not included.
Caution: Warning: Do not heat or subject to pressure. Rated to 1-atmosphere vacuum.