PYREX® Microfiltration All-Glass Assemblies 47 mm, Corning
Supplier: Corning
33971-300 33971-1L 33975-C 33980-300 33980-1L 33982-B 33985-1L 33985-4L
71001-594CS 258.87 CAD
71001-594 71001-596 71001-600 71001-602 71001-604 71001-606 71001-608 71001-610
PYREX® Microfiltration All-Glass Assemblies 47 mm, Corning
Filters Membrane Filter Units
These filtration apparatus and components are designed for HPLC mobile phase filtration, analysis of particulate or bacterial contamination and general laboratory microfiltration. Units have coarse porosity frits, and accommodate 47 mm filters.
  • Chemical compatibility with most organic solvents
Delivery information: The complete system consists of a funnel, flask, 47 mm fritted all-glass support base and aluminium clamp.
Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where suchMore Product Information