Sterile Gauze Sponge, DUKAL™ Corporation
Supplier: Dukal
6208 6312 6408 6842 6208 6312 6408 6842 7070033 7060033 7080033
95041-740CS 189.95 CAD
95041-740 95041-742 95041-744 95041-746 CA95041-740 CA95041-742 CA95041-744 CA95041-746 15431-202 15431-491 15431-500
Sterile Gauze Sponge, DUKAL™ Corporation
First Aid Supplies Wound Dressings
Sterile Gauze Sponges are made of 100% woven cotton.
  • Ideal for a number of applications including general cleaning, dressings, prepping, packing, and debriding wounds
  • Not made with natural rubber latex
Order Now
SPECIFICATIONS
Environmentally Preferable