F1-ClipTip™ Variable Volume Single Channel Pipettes, Thermo Scientific
Supplier: Thermo Scientific
ClipTip™
4641220N 4641210N 4641320N 4641190N 4641230N 4641170N 4641180N 4641200N 4641310N
89348-069EA 803.62 CAD
89348-069 89348-067 76299-428 89348-065 89348-063 76299-426 89348-073 89348-061 89348-071
F1-ClipTip™ Variable Volume Single Channel Pipettes, Thermo Scientific
Pipettes Micropipettes
Now silent and effortless, Thermo Scientific™ F1-ClipTip™ Variable Volume Pipettes feature newly enhanced volume adjustment, and the added confidence of ClipTip technology that locks tips firmly to the pipette with a complete seal.
  • Enhanced volume adjustment
  • Complete seal
  • Remains sealed until ejection
  • High-quality results
  • More efficient research
  • Lightweight design
  • Secure volume lock
With a light touch, each tip is clipped securely on the pipette. When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles.
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI. Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort. The 120° adjustable finger rest, for both right- and left-handed operator comfort, allows the hand to relax during pipetting cycles.
A 150% increase in air boost ensures efficient delivery of micro-volumes and prevent capillary action in 50 μl models and below. Each tip is clipped securely on the pipette. Lifetime manufacturer warranty.
Certifications: The F1-ClipTip pipetting system is CE-marked in accordance with IVD Directive in Europe. Manufactured according to ISO 9001, ISO 14001 and ISO 13485.
Accessories information: Wide selection of ClipTip pipette tips available. Choose from five sizes in 96-format: 20, 50, 200, 300, and 1000 μl.
Ordering information: Industry-leading lifetime warranty with convenient online registration; may vary by country.