Histosette® II Tissue Processing/Embedding Cassettes, Simport
Supplier: Simport Scientific
M485-2 M485-3 M485-5 M485-6 M485-7 M485-8 M485-9 M485-10 M485-11 M485-12 M485-4
CA14230-470CS 333.59 CAD
CA14230-470 CA14230-472 CA14230-476 CA14230-478 CA14230-480 CA14230-482 CA14230-484 CA14230-486 CA14230-488 CA14230-490 CA14230-474
Histosette® II Tissue Processing/Embedding Cassettes, Simport
Embedding Cassettes Tissue Processing Cassettes
These disposable cassettes hold tissue specimens efficiently during the embedding process and in storage
Molded from acetal polymer, they keep specimens submerged in liquid and are resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. Anterior writing area is angled at 45° for suitability with certain types of cassette labeling instruments. Colors are non-cytotoxic and nonmetallic
Ordering information: Lids are supplied unattached. Packed in two dispenser boxes of 500 cassettes and one dispenserMore Product Information