MICROSETTE™ I Biopsy Processing and Embedding Cassettes, Simport Scientific
Supplier: Simport Scientific
M502-2 M502-3 M502-4 M502-5 M502-6 M502-7 M502-8 M502-9 M502-10 M502-11 M502-12 M502-16 M502-17 M502-18
CA48648-311CS 161.79 CAD
CA48648-311 CA48648-312 CA48648-314 CA48648-315 CA48648-316 CA48648-317 CA48648-318 CA48648-319 CA48648-320 CA48648-321 CA48648-322 CA73960-537 CA73960-539 CA73960-541
MICROSETTE™ I Biopsy Processing and Embedding Cassettes, Simport Scientific
Embedding Cassettes Biopsy Processing Cassettes
These innovative disposable plastic cassettes eliminate the need for biopsy pads
Cover and base feature over 2000 square openings, each with a diameter of 0.26mm. These mesh holes allow for maximum fluid exchange. The high-density acetal polymer is resistant to the chemical action of histological solvents. The integral one-piece lids can be opened and closed multiple times. The anterior writing area is set at a 45° angle. Compatible with most cassette printers.
Ordering information: Packaged in dispenser boxes of 150 cassettes with covers assembled, four boxes per case.