SlideFile™ Jr. Storage System, Simport Scientific
Supplier: Simport Scientific
M700-50B M700-50G M700-50P M700-50W M700-50Y
CA48648-520CS 191.59 CAD
CA48648-520 CA48648-522 CA48648-524 CA48648-526 CA48648-528
SlideFile™ Jr. Storage System, Simport Scientific
Slide Holders
Holds up to 200 slides per unit and stackable for space-efficient storage
  • Base, cover, and trays are made of high impact polystyrene
  • Not autoclavable
The slide box includes a removable tray that holds the slides upright. For maximum storage space, remove the tray and line the slides in three rows. A tinted hinged cover makes the contents of the box easy to see.
Temperature range: –80 to 80°C (–112° to 176°F)