SLIMSETTE™ Biopsy Processing and Embedding Cassettes, Simport Scientific
Supplier: Simport Scientific
M510-2 M510-3 M510-4 M510-5 M510-6 M510-7 M510-8 M510-9 M510-10 M510-11 M510-12
CA11031-884CS 433.09 CAD
CA11031-884 CA11031-886 CA11031-888 CA11031-890 CA11031-892 CA11031-894 CA11031-896 CA11031-898 CA11031-900 CA11031-902 CA11031-904
SLIMSETTE™ Biopsy Processing and Embedding Cassettes, Simport Scientific
Embedding Cassettes Biopsy Processing Cassettes
These compact, easy-to-use cassettes incorporate a unique recessed cover, which allows more cassettes to be stacked
They are molded from acetal polymer to keep specimens safely submerged in liquid and are resistant to chemical actions of histological solvents. The 1mm mesh holes are specially designed to hold biopsy specimens, and the one-piece integral lid can be opened and closed as often as necessary. The anterior writing area is slanted at a 45° angle for use with certain types of cassette labeling instruments