ClipTip® Pipette Tips, 100, Thermo Scientific
Supplier: Thermo Fisher Scientific
94410710 94410710 94420718 94410713 94420713 94410717 94410718
89173-450CS 372.95 CAD
89173-450 89348-008 89348-058 89348-018 89348-028 89348-038 89348-048
ClipTip® Pipette Tips, 100, Thermo Scientific
Pipette Tips
Thermo Scientific ClipTip® pipette tips provide security with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force.
  • Certified free of RNase, DNase, DNA, ATP, and pyrogen contamination
  • Suitable for pipetting of 100 to 1000 µl volumes
All ClipTip® pipette tips feature a hydrophobic inner surface which effectively prevent liquid retention. The tips are available in five volumes and in multiple packaging options, including environmentally friendly reload stacks.
The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip® pipette until - and only until - it is released.
Certifications: Manufactured according to ISO 9001 and ISO 13485. The F1-ClipTip® pipetting system is CE-marked in accordance with IVD Directive in Europe.