Corning® Reusable Plastic Volumetric Flasks, PMP, Class A, Corning
Supplier: Corning
5640P-10 5640P-25 5640P-50 5640P-100 5640P-250 5640P-500 5640P-1L
89171-996CS 288.07 CAD
89171-996 89171-998 89172-000 89172-002 89172-004 89172-006 89172-008
Corning® Reusable Plastic Volumetric Flasks, PMP, Class A, Corning
Flasks Volumetric Flasks
PMP, translucent, with tapered PP stopper.
  • Can be autoclaved at 121 ºC without affecting the graduation tolerance
  • Routine cleaning up to 60 ºC will preserve graduation markings
  • With an individually calibrated graduation line
The accuracy of volumetric ware depends on the care used in calibrating it, using the correct type of ware for the application, handling the ware properly and ensuring the ware is clean. Calibration of volumetric ware is usually done at 20 °C, and the ware should be used at approximately this temperature. Refrigerated liquidsMore Product Information