Puritan® Small Rectangular Foam Tipped Applicator, Polypropylene Handle, Sterile, Puritan Medical Products
Supplier: Puritan Medical Products
2516051PSFRE 2516051PSFRE
CA89133-748BX 188.34 CAD
CA89133-748 89133-748
Puritan® Small Rectangular Foam Tipped Applicator, Polypropylene Handle, Sterile, Puritan Medical Products
Swabs Swabs for Sampling
This swab’s small rectangular foam tip has a high collection capacity ideal for diagnostics, forensics, and buccal swabbing as well as industrial cleaning
Made in the USA.
The tip is attached to a rigid polypropylene handle
Certifications: CE marked.
Ordering information: Packaged as 10 boxes of 50 individually wrapped swabs.