Puritan® Oversized Rayon Swab, Proctoscopic Applicator, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
816 816
CA82005-438CS 1472.7 CAD
CA82005-438 82005-438
Puritan® Oversized Rayon Swab, Proctoscopic Applicator, Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Sampling
This swab has an extra-absorbent rayon tip
Autoclavable to 121°C (250°F). Made in USA.
The long polypropylene handle make it ideal for collection in the throat, and for gynecological or proctological exams.
Certifications: CE marked.