Corning® Reusable Plastic Low Form Beakers, PP, Corning
Supplier: Corning
Corning®
1000P-3L 1000P-600 1000P-5L 1000P-50 1000P-250 1000P-1L 1000P-100 1000P-150 1000P-2L 1000P-400 1000P-25
89172-042CS 97.03 CAD
89172-042 89172-054 89172-044 89172-056 89172-050 89172-040 89172-052 89172-046 89172-058 89172-048 89172-038
Corning® Reusable Plastic Low Form Beakers, PP, Corning
Beakers
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable
With printed with blue graduations.