TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Proteomics Grade

Supplier: VWR
Danger

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

M143-1L M143-4L
97063-864EA 84.67 CAD
97063-864 97063-866
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Proteomics Grade
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)

Non-ionic detergent efficiently separates hydrophilic proteins from membrane spanning, hydrophobic proteins without altering biological activity.


  • High purity and protease-free
  • Suitable for most biological applications


These detergents have uncharged hydrophilic groups and are gentle, non-denaturing detergents ideal for studying the conformation and function of proteins, because they keep secondary and tertiary structure intact.

Boiling Pt: 270 °C
Melting Pt: 6
Flash Pt: 251
MDL Number: MFCD00132505
CAS Number: 9002-93-1
Merck Index: 13,06793

Order Now

Specification Test Results

Cloud Point Report
Colour (APHA) ≤100
Peroxides None detectable
pH (5 %, Water) @25 °C 6.0 - 8.0
Protease None detectable


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR