Sodium dodecyl sulfate (SDS), Proteomics Grade
Supplier: VWR
Warning
|
Synonyms:
Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
Negatively charged anionic detergents are strong detergents utilized in lysis buffers for the complete disruption of membranes and denaturation of proteins. Anionic detergents are also used in denaturing gel electrophoresis.
- High purity and protease-free
- Suitable for most biological applications
Formula:
CH₃(CH₂)₁₁OSO₃Na MW: 288.38 g/mol |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 UN: 1325 ADR: 4.1 II Merck Index: 14,08636 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...