Bromophenol blue ACS

Supplier: VWR

Synonyms: Bromphenol Blue Sultone Form, 3,3',5,5'-Tetrabromophenolsulfonphthalein, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein, Tetrabromophenol-sulfonephthalein

0449-50G 0449-25G
97061-692EA 227.84 CAD
97061-692 97061-690
Bromophenol blue ACS
Bromophenol blue
Formula: C₁₉H₁₀Br₄O₅S
MW: 669.97 g/mol
Melting Pt: 273
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2
Merck Index: 13,01429

Order Now

Specification Test Results

Clarity of Solution Pass
Visual Transition Interval (pH 3.0-4.6) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR