γ-(N-Morpholino)propanesulphonic acid (MOPS), Ultrapure

Supplier: VWR
Warning

Synonyms: 3-Morpholinopropane-1-sulfonic acid, 3-(N-Morpholino)propanesulfonic acid, 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid

0670-100G 0670-500G
97062-156EA 136.61 CAD
97062-156 97062-160 97062-158
γ-(N-Morpholino)propanesulphonic acid (MOPS), Ultrapure
γ-(N-Morpholino)propanesulphonic acid (MOPS)

MOPS is a zwitterionic buffer used as a running buffer for denaturing agarose gel electrophoresis of RNA.

Formula: C₇H₁₅NO₄S
MW: 209.27 g/mol
Melting Pt: 277 °C
Flash Pt: 110 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288

Order Now

Specification Test Results

Abs.@260nm (0.1M, Water) = 0.05
Abs.@280nm (0.1M, Water) = 0.05
DNase none detected
RNase none detected
pKa @(25°C) 7 - 7.4
Protease none
Purity 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR